missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ O'RangeRuler DNA Ladders, ready-to-use

Product Code. 10191120 Shop All Thermo Scientific Products
Change view
Click to view available options
Size Range:
10 to 100 bp
10 to 150 bp
100 to 500 bp
100 to 6000 bp
20 to 300 bp
200 to 3000 bp
50 to 1000 bp
500 to 6000 bp
Unit Size:
25µg
50µg
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Range Unit Size
10191120 100 to 500 bp 25µg
10839020 10 to 100 bp 50µg
10765271 10 to 150 bp 50µg
10598170 20 to 300 bp 50µg
10181120 50 to 1000 bp 25µg
10840041 100 to 6000 bp 25µg
10394810 200 to 3000 bp 25µg
10810231 500 to 6000 bp 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10191120 Supplier Thermo Scientific™ Supplier No. SM0623
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific O'RangeRuler DNA Ladders is a step ladder designed for precise sizing of DNA fragments in agarose or non-denaturing polyacrylamide gels.

Thermo Scientific O'RangeRuler DNA Ladders are designed for precise sizing of PCR products and other double-stranded DNA fragments in agarose or non-denaturing polyacrylamide gels.

O'RangeRuler series is characterized by its 'step ladder' design. The chromatography-purified DNA fragments are precisely separated by a consistent increment (e.g., 5 bp, 10 bp, 20 bp, 50 bp, 100 bp, 200 bp, or 500 bp). The design enables accurate and precise sizing of DNA fragments. Not recommended for DNA quantification.

All O'RangeRuler DNA Ladders are supplied with 6X Orange DNA Loading Dye for sample DNA. Orange G dye can be used to monitor DNA migration in agarose or polyacrylamide gels and migrates at 50 bp in a 1% agarose gel.

Highlights

  • Step ladders available in a wide range of increments from 5-to-500 bp
  • Evenly spaced reference bands
  • Sharp bands, easy-to-remember fragment sizes
  • Ready-to-use—premixed with 6X Orange Loading Dye for direct loading
  • Stable at room temperature for up to 24 months

Sustainability features

Ready-to-use O'RangeRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Orange G
Ready to Load Yes
Size Range 100 to 500 bp
Gel Compatibility Agarose Gels
Quantity 25 μg
Product Line O'GeneRuler, O’RangeRuler
Product Type DNA Ladder
Are the Thermo Scientific DNA ladders compatible with GelRed and GelGreen gel stains?

The GelRed or GelGreen nucleic acid dyes can be used with our DNA Ladders. Nucleic acid binding dyes can affect DNA migration during electrophoresis, therefore we highly recommend post-staining of gels. Please follow the gel staining protocol provided by the manufacturer of nucleic acid gel dyes.

Which Thermo Scientific DNA ladders are optimal for fast electrophoresis separation?

For fast sizing and quantification of double-stranded DNA, we recommend using FastRuler DNA Ladders. These ladders are easily resolved in a short separation distance after an 8-14 min run.

What is the recommended voltage for electrophoresis with Thermo Scientific DNA and RNA ladders? What does V/cm mean?

Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).

What are the end functional groups of Thermo Scientific DNA and RNA ladders?

Thermo Scientific DNA and RNA ladders have 5′-phosphate and 3′-hydroxyl groups.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.