missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O,O-Dimethyl Phosphorothionate-d6 Ammonium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | O,O-Dimethyl Phosphorothionate-d6 Ammonium Salt |
| CAS | 1330162-95-2 |
| Molecular Formula | C2H4D6NO3PS |
| Quantity | 5 mg |
| Synonym | Phosphorothioic Acid O,O-Dimethyl Ester-d6 Ammonium Salt, Ammonium Methyl Phosphorothioate-d6, Phosphorothioic Acid O,O-Dimethyl Ester-d6 Ammonium Salt, Ammonium O,O-Dimethyl Phosphorothioate-d6, Ammonium O,O-Dimethyl Thiophosphate-d6, Ammonium Dimethyl Phosphorothioate-d6 |
| InChI Formula | InChI=1S/C2H7O3PS.H3N/c1-4-6(3,7)5-2;/h1-2H3,(H,3,7);1H3/i1D3,2D3; |
| SMILES | SP(OC([2H])([2H])[2H])(OC([2H])([2H])[2H])=O.N |
| Molecular Weight (g/mol) | 165.18 |
| Formula Weight | 165.0496 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?