missing translation for 'onlineSavingsMsg'
Learn More

O,O-Dimethyl Phosphorothionate-13C2 Ammonium Salt, TRC

Product Code. 30386581
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386581 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386581 Supplier Toronto Research Chemicals Supplier No. TRCD4776232.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O,O-Dimethyl Phosphorothionate-13C2 Ammonium Salt
CAS 1330052-30-6
Molecular Formula 13C2H10NO3PS
Quantity 2.5 mg
Synonym Phosphorothioic Acid O,O-Dimethyl Ester-13C2 Ammonium Salt, Ammonium Methyl Phosphorothioate-13C2, Phosphorothioic Acid O,O-Dimethyl Ester-13C2 Ammonium Salt, Ammonium O,O-Dimethyl Phosphorothioate-13C2, Ammonium O,O-Dimethyl Thiophosphate-13C2, Ammonium Dimethyl Phosphorothioate-13C2
InChI Formula InChI=1S/C2H7O3PS.H3N/c1-4-6(3,7)5-2;/h1-2H3,(H,3,7);1H3/i1+1,2+1;
SMILES N.[13CH3]OP(=O)(S)O[13CH3]
IUPAC Name azanium;[(113C)methoxy(sulfido)phosphoryl]oxy(113C)methane
Molecular Weight (g/mol) 161.13
Formula Weight 161.0186
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.