missing translation for 'onlineSavingsMsg'
Learn More

O,O-Dimethyl Dithiophosphate Ammonium Salt, TRC

Product Code. 30386736
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30386736 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386736 Supplier Toronto Research Chemicals Supplier No. TRCD4727401G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O,O-Dimethylphosphorodithioic acid ammonium salt
CAS 1066-97-3
Molecular Formula C2 H6 O2 P S2 . H4 N
Quantity 1 g
Synonym Ammonium methyl phosphorodithioate (6CI,7CI), Phosphorodithioic acid, O,O-dimethyl ester, ammonium salt (8CI,9CI), Ammonium O,O-dimethyl dithiophosphate, Ammonium O,O-dimethyl phosphorodithioate, Ammonium O,O-dimethyl phosphorodithioate(1-), Ammonium O,O'-dimethyl phosphorodithioate, Ammonium dimethyldithiophosphate, O,O-Dimethyl ammonium dithiophosphate, O,O-Dimethyldithiophosphoric acid ammonium salt
InChI Formula InChI=1S/C2H7O2PS2.H3N/c1-3-5(6,7)4-2;/h1-2H3,(H,6,7);1H3
SMILES [NH4+].COP(=S)([S-])OC
IUPAC Name azanium;dimethoxy-sulfanylidene-sulfido-λ^{5}-phosphane
Molecular Weight (g/mol) 175.21
Formula Weight 174.9891
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.