missing translation for 'onlineSavingsMsg'
Learn More

O,O-Diethyl Thiophosphate Potassium Salt, TRC

Product Code. 30387584
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387584 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30387584 Supplier Toronto Research Chemicals Supplier No. TRCD445254500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O,O-Diethyl hydrogen thiophosphate potassium salt
CAS 5871-17-0
Molecular Formula C4 H10 O3 P S . K
Quantity 500 mg
Synonym Phosphorothioic acid, O,O-diethyl ester, potassium salt (1:1), Ethyl potassium phosphorothioate (6CI,7CI), Phosphorothioic acid, O,O-diethyl ester, potassium salt (8CI,9CI), O,O-Diethyl potassium phosphorothioate, Potassium O,O-diethyl phosphorothioate, Potassium O,O-diethylthiophosphate, Potassium di-O-ethyl phosphorothioate, Potassium diethyl thiophosphate
InChI Formula InChI=1S/C4H11O3PS.K/c1-3-6-8(5,9)7-4-2;/h3-4H2,1-2H3,(H,5,9);/q;+1/p-1
SMILES [K+].CCOP(=O)([S-])OCC
IUPAC Name potassium;1-[ethoxy(sulfido)phosphoryl]oxyethane
Molecular Weight (g/mol) 208.26
Formula Weight 207.9725
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.