missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O,O-Diethyl Dithiophosphate, TRC
Specifications
Specifications
| Chemical Name or Material | O,O-Diethyl Dithiophosphate |
| CAS | 298-06-6 |
| Molecular Formula | C4 H11 O2 P S2 |
| Quantity | 500 mg |
| Synonym | Phosphorodithioic acid, O,O-diethyl ester, Di-O-ethyl dithiophosphate, Dithiophosphoric acid O,O-diethyl ester, NSC 171184, O,O-Diethyl dithiophosphate, O,O-Diethyl dithiophosphoric acid ester, O,O-Diethyl hydrogen phosphorodithioate, O,O-Diethyl phosphorodithioate, O,O-Diethyl thiolothionophosphate, O,O'-Diethyl dithiophosphate, O,O'-Diethyl hydrogen dithiophosphate |
| InChI Formula | InChI=1S/C4H11O2PS2/c1-3-5-7(8,9)6-4-2/h3-4H2,1-2H3,(H,8,9) |
| SMILES | CCOP(=S)(S)OCC |
| IUPAC Name | diethoxy-sulfanyl-sulfanylidene-λ^{5}-phosphane |
| Molecular Weight (g/mol) | 186.23 |
| Formula Weight | 185.9938 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?