missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O,O-Diethyl Dithiophosphate-d10 Ammonium salt, TRC
Specifications
Specifications
| Chemical Name or Material | O,O-Diethyl Dithiophosphate-d10 Ammonium Salt |
| Molecular Formula | C4H4D10NO2PS2 |
| Quantity | 50 mg |
| Synonym | Phosphorodithioic Acid O,O-Diethyl Ester-d10 Ammonium Salt, Ammonium Ethyl Phosphorodithioate-d10, Phosphorodithioic Acid O,O-Diethyl Ester-d10 Ammonium Salt, Ammonium O,O-Diethyl Dithiophosphate-d10, Ammonium O,O-Diethyl Phosphorodithioate-d10, Ammonium Di-O-ethyl Dithiophosphate-d10, Ammonium Diethyldithiophosphate-d10, NF 133, O,O-Diethyl Ammonium Phosphorodithioate-d10, O,O-Diethylphosphorodithioic Acid-d10 Ammonium Salt |
| InChI Formula | InChI=1S/C4H11O2PS2.H3N/c1-3-5-7(8,9)6-4-2;/h3-4H2,1-2H3,(H,8,9);1H3/i1D3,2D3,3D2,4D2; |
| SMILES | SP(OC([2H])(C([2H])([2H])[2H])[2H])(OC([2H])(C([2H])([2H])[2H])[2H])=S.N |
| Molecular Weight (g/mol) | 213.32 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?