missing translation for 'onlineSavingsMsg'
Learn More

O,O-Diethyl Dithiophosphate Ammonium Salt, TRC

Product Code. 30379916
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379916 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30379916 Supplier Toronto Research Chemicals Supplier No. TRCD44426525G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O,O-Diethyl Dithiophosphate Ammonium Salt
CAS 1068-22-0
Molecular Formula C4 H10 O2 P S2 . H4 N
Quantity 25 g
Synonym Phosphorodithioic acid, O,O-diethyl ester, ammonium salt (1:1), Ammonium ethyl phosphorodithioate (6CI,7CI), Phosphorodithioic acid, O,O-diethyl ester, ammonium salt (8CI,9CI), Ammonium O,O-diethyl dithiophosphate, Ammonium O,O-diethyl phosphorodithioate, Ammonium O,O-diethyl phosphorodithioate(1-), Ammonium O,O'-diethyldithiophosphate, Ammonium di-O-ethyl dithiophosphate, Ammonium diethyldithiophosphate, Ethyl ammonium phosphorodithioate, (EtO)2(NH4S)PS, Kejunlin, NF 133, O,O-Diethyl ammonium phosphorodithioate, O,O-Diethylphosphorodithioic acid ammonium salt, S-Ammonium O,O'-diethyl phosphorodithioate, O,O-Diethyl dithiophosphate ammonium salt
InChI Formula InChI=1S/C4H11O2PS2.H3N/c1-3-5-7(8,9)6-4-2;/h3-4H2,1-2H3,(H,8,9);1H3
SMILES [NH4+].CCOP(=S)([S-])OCC
IUPAC Name azanium;diethoxy-sulfanylidene-sulfido-λ^{5}-phosphane
Molecular Weight (g/mol) 203.26
Formula Weight 203.0204
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.