missing translation for 'onlineSavingsMsg'
Learn More

O,O-Diethyl Dithiophosphate (>80%), TRC

Product Code. 30380709
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380709 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30380709 Supplier Toronto Research Chemicals Supplier No. TRCD465565250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O,O-Diethyl Dithiophosphate
CAS 298-06-6
Molecular Formula C4 H11 O2 P S2
Quantity 250 mg
Synonym Phosphorodithioic acid, O,O-diethyl ester, Di-O-ethyl dithiophosphate, Dithiophosphoric acid O,O-diethyl ester, NSC 171184, O,O-Diethyl dithiophosphate, O,O-Diethyl dithiophosphoric acid ester, O,O-Diethyl hydrogen phosphorodithioate, O,O-Diethyl phosphorodithioate, O,O-Diethyl thiolothionophosphate, O,O'-Diethyl dithiophosphate, O,O'-Diethyl hydrogen dithiophosphate
InChI Formula InChI=1S/C4H11O2PS2/c1-3-5-7(8,9)6-4-2/h3-4H2,1-2H3,(H,8,9)
SMILES CCOP(=S)(S)OCC
IUPAC Name diethoxy-sulfanyl-sulfanylidene-λ^{5}-phosphane
Molecular Weight (g/mol) 186.23
Formula Weight 185.9938
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.