missing translation for 'onlineSavingsMsg'
Learn More

O,O-Dibutyl Phosphorodithioate Ammonium Salt, TRC

Product Code. 30380003
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380003 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380003 Supplier Toronto Research Chemicals Supplier No. TRCD429380100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O,O-Dibutyl Phosphorodithioate Ammonium Salt
CAS 1071-18-7
Molecular Formula C8H22NO2PS2
Quantity 100 mg
Synonym Butyl Phosphorodithioate Ammonium Salt, O,O-Di-n-Butyl Dithiophosphate Ammonium Salt, O,O-Dibutyl Dithiophosphate Ammonium Salt, O,O-Dibutyl Dithiophosphoric Acid Ammonium Salt, O,O-Dibutyl Hydrogen Phosphorodithioate Ammonium Salt, O,O-Dibutyl Phosphorodithioate Ammonium Salt, O,O'-Dibutyl Dithiophosphate Ammonium Salt, O,O'-Dibutyl Hydrogen Dithiophosphate Ammonium Salt
InChI Formula InChI=1S/C8H19O2PS2.H3N/c1-3-5-7-9-11(12,13)10-8-6-4-2;/h3-8H2,1-2H3,(H,12,13);1H3
SMILES [NH4+].CCCCOP(=S)([S-])OCCCC
IUPAC Name azanium;dibutoxy-sulfanylidene-sulfido-λ^{5}-phosphane
Molecular Weight (g/mol) 259.37
Formula Weight 259.083
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.