missing translation for 'onlineSavingsMsg'
Learn More

(-)-O,O’-Di-p-toluoyl-L-tartaric Acid, TRC

Product Code. 30371636
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371636 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371636 Supplier Toronto Research Chemicals Supplier No. TRCD49438510G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (-)-O,O'-Di-p-toluoyl-L-tartaric Acid
CAS 32634-66-5
Molecular Formula C20 H18 O8
Quantity 10 g
Synonym (2R,3R)-2,3-Bis[(4-methylbenzoyl)oxy]-butanedioic Acid, Butanedioic acid, 2,3-bis[(4-methylbenzoyl)oxy]-, [R-(R*,R*)]-, (-)-Di-O,O' p-toluyltartaric acid, (-)-Di-O,O'-p-toluoyltartaric acid, (-)-Di-O-p-toluoyl-L-tartaric acid, (-)-Di-p-toluoyl-L-tartaric acid, (-)-Di-p-toluoyltartaric acid, (-)-O,O-Di-p-toluoyltartaric acid, (-)-O,O'-Di-p-toluoyl-L-tartaric acid, (2R,3R)-(-)-Di(p-toluoyl)tartaric acid, (2R,3R)-2,3-Bis[(4-methylbenzoyl)oxy]succinic acid, (R,R)-O,O'-Di-p-toluoyltartaric acid, Di-4-toluoyl-L-tartaric acid, Di-p-Toluoyl-d-tartaric acid, Di-p-toluoyl-L-tartaric acid, L-Di-O,O'-p-toluyltartaric acid, L-Di-p-toluoyltartaric acid, O,O'-Di-p-toluoyl-(2R,3R)-tartaric acid, Tartaric acid, di-p-toluate
InChI Formula InChI=1S/C20H18O8/c1-11-3-7-13(8-4-11)19(25)27-15(17(21)22)16(18(23)24)28-20(26)14-9-5-12(2)6-10-14/h3-10,15-16H,1-2H3,(H,21,22)(H,23,24)/t15-,16-/m1/s1
SMILES Cc1ccc(cc1)C(=O)O[C@H]([C@@H](OC(=O)c2ccc(C)cc2)C(=O)O)C(=O)O
IUPAC Name (2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid
Molecular Weight (g/mol) 386.35
Formula Weight 386.1002
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.