missing translation for 'onlineSavingsMsg'
Learn More

(+)-O,O’-Di-p-toluoyl-D-tartaric Acid, TRC

Product Code. 30375574
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30375574 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375574 Supplier Toronto Research Chemicals Supplier No. TRCD4943805G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 32634-68-7
Molecular Formula C20 H18 O8
Quantity 5 g
Synonym (2S,3S)-2,3-Bis[(4-methylbenzoyl)oxy]butanedioic acid, (+)-Di-p-toluoyl-D-(+)-tartaric Acid, Butanedioic acid, 2,3-bis[(4-methylbenzoyl)oxy]-, (2S,3S)-, Butanedioic acid, 2,3-bis[(4-methylbenzoyl)oxy]-, [S-(R*,R*)]-, Tartaric acid, di-p-toluate, (+)- (8CI), p-Toluic acid, diester with D-tartaric acid (7CI), (+)-Bis-O,O'-(4-toluoyl)-D-tartrate, (+)-Di-O,O'-p-toluoyltartaric acid, (+)-Di-O,O'-p-toluyltartaric acid, (+)-Di-O-(p-toluoyl)tartaric acid, (+)-Di-O-p-toluoyl-D-tartaric acid, (+)-Di-p-toluoyl-D-tartaric acid, (+)-Di-p-toluoyltartaric acid, (+)-Ditoluoyltartaric acid, (+)-O,O-Di-p-toluoyltartaric acid, (2S,3S)-(+)-Di-p-toluoyltartaric acid, (2S,3S)-2,3-Bis[(4-methylbenzoyl)oxy]succinic acid, (S,S)-O,O'-Di-p-toluoyltartaric acid, D-Bis(O-4-methylbenzoyl)tartaric acid, D-Di-O,O'-p-toluyltartaric acid, D-Di-p-toluoyltartaric acid, Di-p-toluoyl-D-tartaric acid, Di-p-toluoyl-l-tartaric acid, NSC 97592
InChI Formula InChI=1S/C20H18O8/c1-11-3-7-13(8-4-11)19(25)27-15(17(21)22)16(18(23)24)28-20(26)14-9-5-12(2)6-10-14/h3-10,15-16H,1-2H3,(H,21,22)(H,23,24)/t15-,16-/m0/s1
SMILES Cc1ccc(cc1)C(=O)O[C@@H]([C@H](OC(=O)c2ccc(C)cc2)C(=O)O)C(=O)O
IUPAC Name (2S,3S)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid
Molecular Weight (g/mol) 386.35
Formula Weight 386.1002
Chemical Name or Material (+)-O,O'-Di-p-toluoyl-D-tartaric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.