missing translation for 'onlineSavingsMsg'
Learn More

o-Nitrophenyl Beta-D-Galactopyranoside-6-phosphate, Cyclohexylammonium Salt,, TRC

Product Code. 30481874
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481874 100 mg -
30474279 50 mg -
30473285 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481874 Supplier Toronto Research Chemicals Supplier No. TRCN503625100MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25405-62-3
Molecular Formula C24 H36 N O11 P
Molecular Weight (g/mol) 545.52
IUPAC Name dicyclohexyl [(2R,3R,4S,5R,6S)-3,4,5-trihydroxy-6-(2-nitrophenoxy)oxan-2-yl]methyl phosphate
SMILES O[C@H]1[C@@H](O)[C@@H](COP(=O)(OC2CCCCC2)OC3CCCCC3)O[C@@H](Oc4ccccc4[N+](=O)[O-])[C@@H]1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C24H36NO11P/c26-21-20(34-24(23(28)22(21)27)33-19-14-8-7-13-18(19)25(29)30)15-32-37(31,35-16-9-3-1-4-10-16)36-17-11-5-2-6-12-17/h7-8,13-14,16-17,20-24,26-28H,1-6,9-12,15H2/t20-,21+,22+,23-,24-/m1/s1
Formula Weight 545.2026
Chemical Name or Material o-Nitrophenyl beta-D-Galactopyranoside-6-phosphate Cyclohexylammonium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.