missing translation for 'onlineSavingsMsg'
Learn More

o-Nitroacetophenone, 95%

Product Code. 10113230 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
20 g
Packaging:
Glass bottle
Unit Size:
2.5g
20g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10113230 2.5 g Glass bottle 2.5g
10755231 20 g Glass bottle 20g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10113230 Supplier Thermo Scientific Acros Supplier No. 128320025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 577-59-3 | C8H7NO3 | 165.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-59-3
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00007144
InChI Key SUGXZLKUDLDTKX-UHFFFAOYSA-N
Synonym 2'-nitroacetophenone, 1-2-nitrophenyl ethanone, o-nitroacetophenone, ethanone, 1-2-nitrophenyl, 2-acetylnitrobenzene, acetophenone, 2'-nitro, methyl 2-nitrophenyl ketone, unii-52l53k4x3o, ccris 2329, 1-2-nitrophenyl ethan-1-one
PubChem CID 11346
IUPAC Name 1-(2-nitrophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 28.0°C to 30.0°C
Boiling Point 159.0°C (16.0 mmHg)
Flash Point >112°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4COCH3
Refractive Index 1.5500 to 1.5520
Quantity 2.5 g
Beilstein 07, 288
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether
Formula Weight 165.15
Percent Purity 95%
Chemical Name or Material o-Nitroacetophenone, 95%
Show More Show Less
compliance-icons
Product Identifier
  • o-Nitroacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.