missing translation for 'onlineSavingsMsg'
Learn More

O-Methylisourea hydrochloride, 98%

Product Code. 11437846
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437846 250 g 250g
11417846 10 g 10g
11427846 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437846 Supplier Thermo Scientific Alfa Aesar Supplier No. A14773.30
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5329-33-9 | C2H7ClN2O | 110.541 g/mol

O-methylisourea hydrochloride is used to prepare 1-chloro-2-methyl-isourea. It is also used as an organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-methylisourea hydrochloride is used to prepare 1-chloro-2-methyl-isourea. It is also used as an organic intermediate.

Solubility
Soluble in organic solvents, water, methanol and ethanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5329-33-9
Molecular Formula C2H7ClN2O
Molecular Weight (g/mol) 110.541
MDL Number MFCD00035043
InChI Key MUDVUWOLBJRUGF-UHFFFAOYSA-N
Synonym o-methylisourea hydrochloride, methyl carbamimidate hydrochloride, o-methyluronium chloride, 2-methylpseudourea hydrochloride, o-methyl-isourea hydrochloride, methoxymethanimidamide hydrochloride, carbamimidic acid, methyl ester, monohydrochloride, carbamimidic acid, methyl ester, hydrochloride 1:1, 2-methylpseudourea hcl, acmc-20ak09
PubChem CID 3083899
IUPAC Name methyl carbamimidate;hydrochloride
SMILES COC(=N)N.Cl

Specifications

Melting Point ∼116°C (decomposition)
Quantity 250 g
Beilstein 3909619
Solubility Information Soluble in organic solvents,water,methanol and ethanol.
Formula Weight 110.54
Percent Purity 98%
Chemical Name or Material O-Methylisourea hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.