missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Methylisourea hemisulfate, 94%, Thermo Scientific Chemicals
£52.40 - £690.71
Chemical Identifiers
CAS | 52328-05-9 |
---|---|
Molecular Formula | 0·5 H2SO4 |
Molecular Weight (g/mol) | 123.12 |
MDL Number | MFCD00040594 |
InChI Key | QSCPQKVWSNUJLJ-UHFFFAOYSA-N |
Synonym | o-methylisourea hemisulfate, 2-methylisouronium sulfate, o-methylisourea hemisulfate salt, bis methoxymethanimidamide ; sulfuric acid, carbamimidic acid, methyl ester, sulfate 2:1, omi™, o-methylpseudourea sulfate, 0-methylisourea hemisulfate, o-methylisourea hemisulphate, bis 2-methylisouronium sulphate |
PubChem CID | 11357018 |
IUPAC Name | methyl carbamimidate;sulfuric acid |
SMILES | COC(=N)N.COC(=N)N.OS(=O)(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10591481
|
Thermo Scientific Acros
233030250 |
25 g |
£52.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10347990
|
Thermo Scientific Acros
233032500 |
250 g |
£301.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10423645
|
Thermo Scientific Acros
233030010 |
1 kg | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
52328-05-9 | |
123.12 | |
QSCPQKVWSNUJLJ-UHFFFAOYSA-N | |
11357018 | |
COC(=N)N.COC(=N)N.OS(=O)(=O)O |
0·5 H2SO4 | |
MFCD00040594 | |
o-methylisourea hemisulfate, 2-methylisouronium sulfate, o-methylisourea hemisulfate salt, bis methoxymethanimidamide ; sulfuric acid, carbamimidic acid, methyl ester, sulfate 2:1, omi™, o-methylpseudourea sulfate, 0-methylisourea hemisulfate, o-methylisourea hemisulphate, bis 2-methylisouronium sulphate | |
methyl carbamimidate;sulfuric acid |
Specifications
52328-05-9 | |
100.0 | |
94% | |
0·5 H2SO4 | |
MFCD00040594 | |
Solubility in water: 1000g/L water (20°C) | |
COC(=N)N.COC(=N)N.OS(=O)(=O)O | |
123.12 | |
123.12 | |
O-Methylisourea hemisulfate, 94% |
92.0 | |
147°C to 153°C | |
Plastic bottle | |
0·5H2SO4 | |
o-methylisourea hemisulfate, 2-methylisouronium sulfate, o-methylisourea hemisulfate salt, bis methoxymethanimidamide ; sulfuric acid, carbamimidic acid, methyl ester, sulfate 2:1, omi™, o-methylpseudourea sulfate, 0-methylisourea hemisulfate, o-methylisourea hemisulphate, bis 2-methylisouronium sulphate | |
QSCPQKVWSNUJLJ-UHFFFAOYSA-N | |
methyl carbamimidate;sulfuric acid | |
11357018 | |
94% |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 257-851-8
RUO – Research Use Only