missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals O-Methyl-L-tyrosine, 98%

Product Code. 10167703 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10167703 5 g Glass Bottle 5g
10433035 250 mg Glass Bottle 250mg
10042150 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10167703 Supplier Thermo Scientific Chemicals Supplier No. 169750050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6230-11-1 | C10H13NO3 | 195.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6230-11-1
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00002604
InChI Key GEYBMYRBIABFTA-IMWMWJONNA-N
Synonym 4-methoxy-l-phenylalanine, o-methyl-l-tyrosine, h-tyr me-oh, o-methyltyrosine, s-2-amino-3-4-methoxyphenyl propanoic acid, p-methoxy-l-phenylalanine, 2s-2-amino-3-4-methoxyphenyl propanoic acid, tyrosine, o-methyl, l-4-methoxyphenylalanine, h-4-methoxy-phe-oh
PubChem CID 2723935
IUPAC Name (2S)-2-amino-3-(4-methoxyphenyl)propanoic acid
SMILES COC1=CC=C(C[C@H](N)C(O)=O)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CH3OC6H4CH2CH(NH2)CO2H
Beilstein 14, I, 668
Specific Rotation -7° to -9° (20°C, 589nm) (c=2, 1N HCl)
Specific Rotation Condition −8° (20°C c=2,1N HCl)
Quantity 5 g
Formula Weight 195.22
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material O-Methyl-L-tyrosine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.