missing translation for 'onlineSavingsMsg'
Learn More

o,m,p-Isopropylphenyl Phenyl Phosphate Mixture (1:1:1), TRC

Product Code. 30397749
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397749 25 mg
30408101 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397749 Supplier Toronto Research Chemicals Supplier No. TRCI87302025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68782-95-6
Molecular Formula C12 H11 O4 P . C4 H10
Molecular Weight (g/mol) 308.309
IUPAC Name diphenyl hydrogen phosphate;2-methylpropane
SMILES CC(C)C.OP(=O)(Oc1ccccc1)Oc2ccccc2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H11O4P.C4H10/c13-17(14,15-11-7-3-1-4-8-11)16-12-9-5-2-6-10-12;1-4(2)3/h1-10H,(H,13,14);4H,1-3H3
Formula Weight 308.118
Chemical Name or Material o,m,p-Isopropylphenyl Phenyl Phosphate Mixture (1:1:1)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.