Learn More
Invitrogen™ O-linked N-acetylglucosamine (O-GlcNAc) Monoclonal Antibody (HGAC85)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA1076
Description
MA1-076 recognizes beta-1,3 linked O-linked N-acetylglucosamine (O-GlcNAc) residues of streptococcal group A carbohydrate as well as O-GlcNAc glycosylated proteins. MA1-076 has been successfully used in Western blot, immunofluorescence, immunoprecipitation and ELISA procedures. By Western blot, this antibody detects several proteins representing immunoprecipitated O-GlcNAc glycoproteins. Immunofluorescence staining of O-GlcNAc in cells results in labeling of the nuclear envelope and pores, nucleolus, and cytoplasm. This staining pattern is consistent with other methods of detecting O-GlcNAc moieties. The MA1-076 immunogen is heat killed, pepsin treated group A streptococci. DO NOT USE WITH DILUENTS CONTAINING GLYCOSYLATED PROTEINS.
O-linked N-acetylglucosamine (O-GlcNAc) is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine or threonine residues. Unlike other protein glycosylations, O-GlcNAc modifications occur within the nucleus and cytoplasm. They are found on many cellular proteins, including nuclear pore, oncogene, cytoskeletal, heat shock, viral and transcription regulatory proteins. O-GlcNAc glycosylations are thought to obscure phosphorylation sites, counteracting phosphorylation-dependent signaling pathways and protein interactions.
Specifications
| O-linked N-acetylglucosamine (O-GlcNAc) | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites, PBS with 0.05% sodium azide | |
| O15294, Q8CGY8, P56558 | |
| OGT | |
| Heat killed, pepsin treated group A streptococci. | |
| RUO | |
| 108155, 26295, 8473 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| HGAC85 | |
| Unconjugated | |
| OGT | |
| GlcNAc; HINCUT-1; HRNT1; O-GLCNAC; O-GlcNAc transferase p110 subunit; O-GlcNAc transferase subunit p110; OGT; O-linked N-acetylglucosamine (GlcNAc) transferase; O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase); O-linked N-acetylglucosamine transferase 110 kDa subunit; UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase; UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDa subunit; uridinediphospho-N-acetylglucosamine:polypeptide beta-N-acetylglucosaminyl transferase | |
| Mouse | |
| 100 μL | |
| Primary | |
| Chemical | |
| Antibody | |
| IgG3 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.