missing translation for 'onlineSavingsMsg'
Learn More

O-Isopropyl (5-Bromoquinoxalin-6-yl)carbamothioate(Brimonidine Impurity), TRC

Product Code. 30394745
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394745 250 mg 250mg
30397090 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394745 Supplier Toronto Research Chemicals Supplier No. TRCI824230250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O-Isopropyl (5-Bromoquinoxalin-6-yl)carbamothioate(Brimonidine Impurity)
Analyte or Component Names O-Isopropyl (5-Bromoquinoxalin-6-yl)carbamothioate(Brimonidine Impurity)
Purity Grade Notes HPLC
Shipping Condition Room Temperature
Molecular Formula C12H12BrN3OS
InChI Formula InChI=1S/C12H12BrN3OS/c1-7(2)17-12(18)16-8-3-4-9-11(10(8)13)15-6-5-14-9/h3-7H,1-2H3,(H,16,18)
Solution Type Neat
SMILES CC(C)OC(=S)Nc1ccc2nccnc2c1Br
Industry Type Pharmaceutical
Molecular Weight (g/mol) 326.21
Quantity 250 mg
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.