missing translation for 'onlineSavingsMsg'
Learn More

o-Iodohippuric Acid, TRC

Product Code. 30406844
Change view
Click to view available options
Quantity:
1 g
2.5 g
10 g
Unit Size:
10g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406844 1 g -
30395604 10 g 10g
30405649 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406844 Supplier Toronto Research Chemicals Supplier No. TRCI7065401G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-58-0
Molecular Formula C9 H8 I N O3
Molecular Weight (g/mol) 305.07
Synonym Glycine, N-(2-iodobenzoyl)-, Hippuric acid, o-iodo- (6CI,8CI), N-(2-Iodobenzoyl)glycine, 2-Iodohippuric acid, N-(o-Iodobenzoyl)glycine, o-Iodohippuric acid
IUPAC Name 2-[(2-iodobenzoyl)amino]acetic acid
SMILES OC(=O)CNC(=O)c1ccccc1I

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H8INO3/c10-7-4-2-1-3-6(7)9(14)11-5-8(12)13/h1-4H,5H2,(H,11,14)(H,12,13)
Formula Weight 304.9549
Chemical Name or Material o-Iodohippuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.