missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
O-GlcNAcase/OGA/MGEA5 Polyclonal antibody specifically detects O-GlcNAcase/OGA/MGEA5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Applications | Western Blot, Immunohistochemistry |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 500 to 550 of human Meningioma Expressed Antigen 5 using the numbering given in entry NP_036347.1 (GeneID 10724). |
| Purification Method | Immunogen affinity purified |
| Gene ID (Entrez) | 10724 |
| Content And Storage | Store at 4°C in the dark. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?