missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-GlcNAcase/OGA/MGEA5 Antibody (9B9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42998
This item is not returnable.
View return policy
Description
O-GlcNAcase/OGA/MGEA5 Monoclonal antibody specifically detects O-GlcNAcase/OGA/MGEA5 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| O-GlcNAcase/OGA/MGEA5 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 10724 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 9B9 | |
| Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/ Immunofluorescence 1:20 - 1:200 | |
| bifunctional protein NCOAT, FLJ11229, HEXC, hyaluronidase in meningioma, KIAA0679, MEA5FLJ23355, meningioma expressed antigen 5 (hyaluronidase), Meningioma-expressed antigen 5, NCOAT, Nuclear cytoplasmic O-GlcNAcase and acetyltransferase, OGA, O-GlcNAcase | |
| meningioma expressed antigen 5(hyaluronidase) - (Uniprot# O60502) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction