missing translation for 'onlineSavingsMsg'
Learn More

O-GlcNAc Transferase p110 subunit Rabbit anti-Human, Mouse, Rat, Clone: 3Q9B8, Novus Biologicals™

Product Code. 18358045 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358045 100 μg 100µL
18376134 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18358045 Supplier Novus Biologicals Supplier No. NBP316203100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

O-GlcNAc Transferase p110 subunit Monoclonal antibody specifically detects O-GlcNAc Transferase p110 subunit in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen O-GlcNAc Transferase p110 subunit
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 3Q9B8
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 2.4.1, EC 2.4.1.255, FLJ23071, HRNT1, MGC22921, O-GLCNAC, O-GlcNAc transferase p110 subunit, O-GlcNAc transferase subunit p110, O-linked N-acetylglucosamine (GlcNAc) transferase(UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase), O-linked N-acetylglucosamine transferase 110 kDa subunit, UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDasubunit, uridinediphospho-N-acetylglucosamine:polypeptide beta-N-acetylglucosaminyltransferase
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 947-1046 of human O-GlcNAc Transferase p110 subunit (O15294).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8473
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.