missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-GlcNAc Antibody (9B9), Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £504.00
Specifications
| Antigen | O-GlcNAc |
|---|---|
| Clone | 9B9 |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18247121
|
Novus Biologicals
NBP2-59309 |
100 μg |
£504.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18644107
|
Novus Biologicals
NBP2-59309-25ug |
25 μg |
£150.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
O-GlcNAc Monoclonal specifically detects O-GlcNAc in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| O-GlcNAc | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Synthetic B-O-Linked N-Acetylglucosamine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| Primary | |
| Specific for B-O-Linked N-Acetylgalactosamine and B-O-Linked N-Acetylglucosamine modified proteins. Does not cross-react with Glucose or Galactose modified proteins. |
| 9B9 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| GlcNAc, O-linked N-acetylglucosamine | |
| IgM | |
| PEG purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title