missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-GalNAc, Mouse anti-Human, Biotin, Clone: 9B9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB16477.100ug
This item is not returnable.
View return policy
Specifications
| O-GalNAc | |
| Monoclonal | |
| Biotin | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
| Synthetic β-O-Linked N-Acetylglucosamine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| RUO | |
| Human | |
| Antibody | |
| IgM |
| ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
| 9B9 | |
| ELISA (1:1000) Immunocytochemistry (1:50) Immunofluorescence (1:50) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction