missing translation for 'onlineSavingsMsg'
Learn More

O-Ethyl O-Phenyl S-Propyl Phosphorothioate, TRC

Product Code. 30382352
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382352 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382352 Supplier Toronto Research Chemicals Supplier No. TRCE9167455MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Heterophos
CAS 40626-35-5
Molecular Formula C11 H17 O3 P S
Quantity 5 mg
Synonym Fostil, Heterofos, Heterophos, O-Ethyl O-phenyl S-propyl phosphorothioate, O-Ethyl O-phenyl S-propyl thiophosphate, Phostil
InChI Formula InChI=1S/C11H17O3PS/c1-3-10-16-15(12,13-4-2)14-11-8-6-5-7-9-11/h5-9H,3-4,10H2,1-2H3
SMILES CCCSP(=O)(OCC)Oc1ccccc1
IUPAC Name [ethoxy(propylsulfanyl)phosphoryl]oxybenzene
Molecular Weight (g/mol) 260.29
Formula Weight 260.064
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.