missing translation for 'onlineSavingsMsg'
Learn More

O-Ethyl Albuterol, TRC

Product Code. 30380276
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380276 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380276 Supplier Toronto Research Chemicals Supplier No. TRCE89750025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-[{(1,1-Dimethylethyl)amino}methyl]-3-(ethoxy-methyl)-4-hydroxy-benzenemethanol
CAS 754926-25-5
Molecular Formula C15 H25 N O3
Quantity 25 mg
Synonym Benzenemethanol, α-[[(1,1-dimethylethyl)amino]methyl]-3-(ethoxymethyl)-4-hydroxy-, BIIK 0277, Levalbuterol Related Compound E (USP), (1RS)-2-[(1,1-Dimethylethyl)amino]-1-[4-hydroxy-3-(ethoxymethyl)phenyl]ethanol, O-Ethyl Albuterol, GW 403983X, Ph Eur Salbutamol Impurity O, Salbutamol ethyl ether
InChI Formula InChI=1S/C15H25NO3/c1-5-19-10-12-8-11(6-7-13(12)17)14(18)9-16-15(2,3)4/h6-8,14,16-18H,5,9-10H2,1-4H3
SMILES CCOCc1cc(ccc1O)C(O)CNC(C)(C)C
IUPAC Name 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(ethoxymethyl)phenol
Molecular Weight (g/mol) 267.36
Formula Weight 267.1834
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.