missing translation for 'onlineSavingsMsg'
Learn More

O-((Ethoxycarbonyl)cyanomethyleneamino)-N,N,N',N'-tetramethyluronium BF4, 98%, ACROS Organics™

Product Code. 10752623 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10752623 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10752623 Supplier Thermo Scientific Acros Supplier No. 310010250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Coupling reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 136849-72-4
CAS Min % 97.5
CAS Max % 100.0
Melting Point 141°C to 147°C
Color White to Yellow
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H17BF4N4O3
MDL Number MFCD00192127 MFCD00192127
Quantity 25 g
Synonym totu, o-cyano ethoxycarbonyl methylenamino-1,1,3,3-tetramethyluronium tetrafluoroborate, o-ethoxycarbonyl cyanomethylenamino n,n,n',n'-tetramethyluronium tetrafluoroborate, o-ethoxycarbonyl cyanomethylenamino-n,n,n',n'-tetra methyluronium tetrafluoroborate, o-ethoxycarbonyl cyanomethyleneamino-n,n,n',n'-tetramethyluronium bf4, e-2-1-cyano-2-ethoxy-2-oxoethylideneamino-1,1,3,3-tetramethyluronium tetrafluoroborate, o-ethoxycarbonyl cyanomethylenamino-n,n,n'n'-tetramethyluronium tetrafluoroborate, o-ethoxycarbonyl cyanomethyleneamino-n,n,n',n'-tetramethyl uronium tetrafluoroborate, o-ethoxycarboxyl cyanomethyleneamino-n,n,n',n'-tetramethyluronium tetrafluoroborate
InChI Key FPQVGDGSRVMNMR-UHFFFAOYSA-N
SMILES F[B-](F)(F)F.CCOC(=O)C(=NOC(N(C)C)=[N+](C)C)C#N
IUPAC Name ethyl cyano({[(dimethylamino)(dimethyliminiumyl)methoxy]imino})formate; tetrafluoroboranuide
Molecular Weight (g/mol) 328.07
PubChem CID 9601231
Formula Weight 327.95
Physical Form Crystalline Powder
Chemical Name or Material O-((Ethoxycarbonyl)cyanomethyleneamino)-N,N,N';,N';-tetramethyluronium BF4, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.