missing translation for 'onlineSavingsMsg'
Learn More

O-Dibenzo[b,f][1,4]thiazepinyl Quetiapine, TRC

Product Code. 30370002
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370002 5 mg
30383211 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30370002 Supplier Toronto Research Chemicals Supplier No. TRCE6783705MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1800608-95-0
Molecular Formula C34 H32 N4 O2 S2
Molecular Weight (g/mol) 592.7735
Synonym Dibenzo[b,f][1,4]thiazepine, 11-[4-[2-[2-(dibenzo[b,f][1,4]thiazepin-11-yloxy)ethoxy]ethyl]-1-piperazinyl]-, Quetiapine Fumarate Imp. W (EP)
IUPAC Name 6-[4-[2-(2-benzo[b][1,4]benzothiazepin-6-yloxyethoxy)ethyl]piperazin-1-yl]benzo[b][1,4]benzothiazepine
SMILES C(COC1=Nc2ccccc2Sc3ccccc13)OCCN4CCN(CC4)C5=Nc6ccccc6Sc7ccccc57

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C34H32N4O2S2/c1-5-13-29-25(9-1)33(35-27-11-3-7-15-31(27)41-29)38-19-17-37(18-20-38)21-22-39-23-24-40-34-26-10-2-6-14-30(26)42-32-16-8-4-12-28(32)36-34/h1-16H,17-24H2
Formula Weight 592.1967
Chemical Name or Material 11-(4-[2-[2-(Dibenzo[b,f][1,4]thiazepin-11-yloxy)ethoxy]ethyl]piperazin-1-yl)dibenzo[b,f][1,4]thiazepine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.