missing translation for 'onlineSavingsMsg'
Learn More

o-Dianisidine, TRC

Product Code. 18090440
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18090440 2.5 g 2.5g
18010350 500 mg 500mg
18053550 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18090440 Supplier Toronto Research Chemicals Supplier No. TRCD4164982.5G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-90-4
Molecular Formula C14 H16 N2 O2
Molecular Weight (g/mol) 244.29
Synonym Benzidine, 3,3'-dimethoxy- (8CI), Fast Blue B Base (6CI), 3,3'-Dimethoxy[1,1'-biphenyl]-4,4'-diamine, 3,3'-Dimethoxy-4,4'-diaminobiphenyl, 3,3'-Dimethoxy-4,4'-diaminodiphenyl, 3,3'-Dimethoxybenzidene, 3,3'-Dimethoxybenzidine, 4,4'-Bi-o-anisidine, 4,4'-Diamino-3,3'-dimethoxy-1,1'-biphenyl, Amacel Developed Navy SD, Azogene Fast Blue B, Blue BN Base, Blue Base Irga B, Blue Base NB, C.I. Disperse Black 6, Cellitazol B, Cibacete Diazo Navy Blue 2B, Diacel Navy DC, Dianisidine, Fast Blue Base B, Fast Blue DSC Base, Hiltonil Fast Blue B Base, Kayaku Blue B Base, Lake Blue B Base, Mitsui Blue B Base, NSC 3168, Naphthanil Blue B Base, Setacyl Diazo Navy R, o-Dianisidine
IUPAC Name 4-(4-amino-3-methoxyphenyl)-2-methoxyaniline
SMILES COc1cc(ccc1N)c2ccc(N)c(OC)c2

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C14H16N2O2/c1-17-13-7-9(3-5-11(13)15)10-4-6-12(16)14(8-10)18-2/h3-8H,15-16H2,1-2H3
Formula Weight 244.1212
Chemical Name or Material 3,3'-Dimethoxybenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.