missing translation for 'onlineSavingsMsg'
Learn More

o-Dianisidine, free base, 99.9%, MP Biomedicals™

Product Code. 11417890 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417890 1 g 1g
11437890 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417890 Supplier MP Biomedicals Supplier No. 0210153601
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 119-90-4
Molecular Formula C14H16N2O2
Molecular Weight (g/mol) 244.294
InChI Key JRBJSXQPQWSCCF-UHFFFAOYSA-N
Synonym 3,3'-dimethoxybenzidine, o-dianisidine, dianisidine, cellitazol b, neutrosel navy bn, diacel navy dc, disperse black 6, blue base nb, blue bn base, blue base irga b
PubChem CID 8411
ChEBI CHEBI:82321
IUPAC Name 4-(4-amino-3-methoxyphenyl)-2-methoxyaniline
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)OC)N

Specifications

Color Brown
Assay Percent Range 99.9%
Quantity 1 g
Formula Weight 244.3
Physical Form Powder
Chemical Name or Material o-Dianisidine, free base
compliance-icons compliance-icons
Product Identifier
  • o-Dianisidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
Hazard Statement
  • H302-Harmful if swallowed.
  • H350-May cause cancer.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 3,3'-dimethoxybenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.