missing translation for 'onlineSavingsMsg'
Learn More

O-Desmorpholinopropyl Gefitinib, TRC

Product Code. 30136280
Change view
Click to view available options
Quantity:
100 mg
1 g
25 g
Unit Size:
100mg
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136280 100 mg 100mg
30134624 25 g 25g
30009612 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136280 Supplier Toronto Research Chemicals Supplier No. TRCD296830100MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 184475-71-6
Molecular Formula C15 H11 Cl F N3 O2
Molecular Weight (g/mol) 319.72
Synonym 6-Quinazolinol, 4-[(3-chloro-4-fluorophenyl)amino]-7-methoxy-, 4-[(3-Chloro-4-fluorophenyl)amino]-7-methoxy-6-quinazolinol, 4-(3-Chloro-4-fluoroanilino)-6-hydroxy-7-methoxyquinazoline, 4-(3-Chloro-4-fluorophenylamino)-6-hydroxy-7-methoxyquinazoline, 4-(3-Chloro-4-fluorophenylamino)-7-methoxyquinazolin-6-ol, N-(3-Chloro-4-fluorophenyl)-6-hydroxy-7-methoxyquinazolin-4-amine, O-Desmorpholinopropyl Gefitinib
IUPAC Name 4-(3-chloro-4-fluoroanilino)-7-methoxyquinazolin-6-ol
SMILES COc1cc2ncnc(Nc3ccc(F)c(Cl)c3)c2cc1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H11ClFN3O2/c1-22-14-6-12-9(5-13(14)21)15(19-7-18-12)20-8-2-3-11(17)10(16)4-8/h2-7,21H,1H3,(H,18,19,20)
Formula Weight 319.0524
Chemical Name or Material O-Desmorpholinopropyl Gefitinib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.