missing translation for 'onlineSavingsMsg'
Learn More

O-Desmethyl O-TBDMS Primaquine, TRC

Product Code. 30357745
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357745 250 mg 250mg
30354228 1 g 1g
30354925 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357745 Supplier Toronto Research Chemicals Supplier No. TRCD291558250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O-Desmethyl O-TBDMS Primaquine
Analyte or Component Names O-Desmethyl O-TBDMS Primaquine
Shipping Condition Room Temperature
Molecular Formula C20 H33 N3 O Si
Synonym N4-(6-((tert-Butyldimethylsilyl)oxy)quinolin-8-yl)pentane-1,4-diamine
InChI Formula InChI=1S/C20H33N3OSi/c1-15(9-7-11-21)23-18-14-17(24-25(5,6)20(2,3)4)13-16-10-8-12-22-19(16)18/h8,10,12-15,23H,7,9,11,21H2,1-6H3
Solution Type Neat
SMILES CC(CCCN)Nc1cc(O[Si](C)(C)C(C)(C)C)cc2cccnc12
IUPAC Name 4-N-[6-[tert-butyl(dimethyl)silyl]oxyquinolin-8-yl]pentane-1,4-diamine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 359.581
Quantity 250 mg
Formula Weight 359.239
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.