missing translation for 'onlineSavingsMsg'
Learn More

O-Desmethyl Gefitinib, TRC

Product Code. 30137025
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137025 5 mg 5mg
30133639 10 mg 10mg
17904405 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137025 Supplier Toronto Research Chemicals Supplier No. TRCD2916805MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 847949-49-9
Molecular Formula C21 H22 Cl F N4 O3
Molecular Weight (g/mol) 432.88
Synonym 7-Quinazolinol, 4-[(3-chloro-4-fluorophenyl)amino]-6-[3-(4-morpholinyl)propoxy]-, 4-[(3-Chloro-4-fluorophenyl)amino]-6-[3-(4-morpholinyl)propoxy]-7-quinazolinol, N-(3-Chloro-4-fluorophenyl)-7-hydroxy-6-[3-(morpholin-4-yl)propoxy]quinazolin-4-amine, 4-[(3-Chloro-4-fluorophenyl)amino]-6-[3-(morpholin-4-yl)propoxy]quinazolin-7-ol
IUPAC Name 4-(3-chloro-4-fluoroanilino)-6-(3-morpholin-4-ylpropoxy)quinazolin-7-ol
SMILES Oc1cc2ncnc(Nc3ccc(F)c(Cl)c3)c2cc1OCCCN4CCOCC4

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H22ClFN4O3/c22-16-10-14(2-3-17(16)23)26-21-15-11-20(19(28)12-18(15)24-13-25-21)30-7-1-4-27-5-8-29-9-6-27/h2-3,10-13,28H,1,4-9H2,(H,24,25,26)
Formula Weight 432.1364
Chemical Name or Material O-Desmethyl Gefitinib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.