missing translation for 'onlineSavingsMsg'
Learn More

O-Desethyl Dapagliflozin tetraacetate), TRC

Product Code. 30373842
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30373842 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373842 Supplier Toronto Research Chemicals Supplier No. TRCE521490250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Des(tetrahydrofuran) Empagliflozin Tetraacetate
CAS 1079083-63-8
Molecular Formula C27H29ClO10
Quantity 250 mg
Synonym Des(tetrahydrofuran) Empagliflozin Tetraacetate, (1S)-1,5-anhydro-1-C-[4-chloro-3-[(4-hydroxyphenyl)methyl]phenyl]-, 2,3,4,6-tetraacetate D-Glucitol, (2R,3R,4R,5S,6S)-2-(Acetoxymethyl)-6-(4-chloro-3-(4-hydroxybenzyl)phenyl)tetrahydro-2H-pyran-3,4,5-triyl Triacetate
InChI Formula InChI=1S/C27H29ClO10/c1-14(29)34-13-23-25(35-15(2)30)27(37-17(4)32)26(36-16(3)31)24(38-23)19-7-10-22(28)20(12-19)11-18-5-8-21(33)9-6-18/h5-10,12,23-27,33H,11,13H2,1-4H3/t23-,24+,25-,26+,27+/m1/s1
SMILES CC(=O)OC[C@H]1O[C@H]([C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C)c2ccc(Cl)c(Cc3ccc(O)cc3)c2
IUPAC Name (2R,3R,4R,5S,6S)-2-(acetoxymethyl)-6-(4-chloro-3-(4-hydroxybenzyl)phenyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate
Molecular Weight (g/mol) 548.97
Formula Weight 548.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.