missing translation for 'onlineSavingsMsg'
Learn More

O-De-phenyl Andarine, TRC

Product Code. 30137947
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137947 100 mg 100mg
30134894 250 mg 250mg
30012406 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137947 Supplier Toronto Research Chemicals Supplier No. TRCD221630100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1022087-30-4
Molecular Formula C11 H11 F3 N2 O5
Molecular Weight (g/mol) 308.211
Synonym Propanamide, 2,3-dihydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]-, (2S)- (ACI), (2S)-2,3-Dihydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide (ACI), O-De-phenyl Andarine
IUPAC Name (2S)-2,3-dihydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
SMILES C[C@](O)(CO)C(=O)Nc1ccc(c(c1)C(F)(F)F)[N+](=O)[O-]

Especificaciones

Quantity 100 mg
InChI Formula InChI=1S/C11H11F3N2O5/c1-10(19,5-17)9(18)15-6-2-3-8(16(20)21)7(4-6)11(12,13)14/h2-4,17,19H,5H2,1H3,(H,15,18)/t10-/m0/s1
Formula Weight 308.062
Chemical Name or Material O-De-phenyl Andarine
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.