missing translation for 'onlineSavingsMsg'
Learn More

o-Cresolphthalein Complexone Sodium Salt, Thermo Scientific Chemicals

Product Code. 10599272 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10599272 25 g Glass bottle 25g
10127180 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10599272 Supplier Thermo Scientific Acros Supplier No. 405750250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62698-54-8
Molecular Formula C32H28N2Na4O12
Molecular Weight (g/mol) 724.54
MDL Number MFCD00066371
InChI Key KSMPPVYJZJBIES-UHFFFAOYSA-J
PubChem CID 112916
IUPAC Name tetrasodium;2-[[5-[1-[3-[[bis(carboxylatomethyl)amino]methyl]-4-hydroxy-5-methylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl]methyl-(carboxylatomethyl)amino]acetate
SMILES [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1

Specifications

Infrared Spectrum Authentic
Formula Weight 724.53
Grade Extra Pure
Packaging Glass bottle
Suitability (for use as a reagent for calcium) passes
Color Purple
Quantity 25 g
Physical Form Powder
Chemical Name or Material o-Cresolphthalein complexone sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.