missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals o-Cresolphthalein complexone, indicator grade

Product Code. 11306686
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11306686 25 g 25g
11386676 1 g 1g
11396676 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11306686 Supplier Thermo Scientific Chemicals Supplier No. L03859.14
  • ONLINE EXCLUSIVE PRICE
    £215.22
    £175.65 / 25g
    Save £39.57
    18% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2411-89-4 | C32H32N2O12 | 636.61 g/mol

Indicator for complexometric titration of Mg, Ca, Sr and Ba. It is used as an alkaline earth metal ion indicator. Suggested to be colorless at pH 11, but turns pink in the presence of alkaline earth metals. Also utilized in an auto analyzer to determine serum calcium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indicator for complexometric titration of Mg, Ca, Sr and Ba. It is used as an alkaline earth metal ion indicator. Suggested to be colorless at pH 11, but turns pink in the presence of alkaline earth metals. Also utilized in an auto analyzer to determine serum calcium.

Solubility
Soluble in NaOH (50 mg/ml), aqueous ammonia, and organic solvents. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2411-89-4
Molecular Formula C32H32N2O12
Molecular Weight (g/mol) 636.61
MDL Number MFCD00005911
InChI Key IYZPEGVSBUNMBE-UHFFFAOYSA-N
Synonym o-cresolphthalein complexone, phthalein purple, cresolphthalexon, o-cresolphthalexon, phthalein complexon, o-cresolphthalein complexon, cresolphthalein complexon, metal phthalein, cresolphthalein complexone, unii-a4p6737i7f
PubChem CID 75485
IUPAC Name 2-[[5-[1-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-5-methylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl]methyl-(carboxymethyl)amino]acetic acid
SMILES CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1

Specifications

Melting Point ∽198°C (decomposition)
Quantity 25 g
Beilstein 381994
Solubility Information Soluble in NaOH (50mg/ml),aqueous ammonia,and organic solvents. Insoluble in water.
Formula Weight 636.62
Grade Indicator
Chemical Name or Material o-Cresolphthalein complexone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.