missing translation for 'onlineSavingsMsg'
Learn More

o-Chloranil, TRC

Product Code. 30355315
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30355315 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30355315 Supplier Toronto Research Chemicals Supplier No. TRCC3252451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material o-Chloranil
CAS 2435-53-2
Molecular Formula C6Cl4O2
Quantity 1 g
Synonym 3,4,5,6-Tetrachloro-3,5-cyclohexadiene-1,2-dione, 3,4,5,6-Tetrachloro-o-benzoquinone, Tetrachloro-o-benzoquinone, 3,4,5,6-Tetrachloro-1,2-benzoquinone, 3,4,5,6-Tetrachlorocyclohexadiene-1,2-dione, Isochloranil, NSC 403503, Tetrachloro-1,2-benzoquinone, Tetrachloro-o-benzoquinone, Tetrachloro-o-quinone, Tetrachloro-ortho-benzoquinone, Tetrachloroorthobenzoquinone
InChI Formula InChI=1S/C6Cl4O2/c7-1-2(8)4(10)6(12)5(11)3(1)9
SMILES ClC1=C(Cl)C(=O)C(=O)C(=C1Cl)Cl
IUPAC Name 3,4,5,6-tetrachlorocyclohexa-3,5-diene-1,2-dione
Molecular Weight (g/mol) 245.88
Formula Weight 243.8652
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.