missing translation for 'onlineSavingsMsg'
Learn More

O-Benzylphospho-N-Fmoc-L-serine, 95%

Product Code. 15438707
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438707 5 g 5g
15448707 250 mg 250mg
15428707 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15438707 Supplier Thermo Scientific Alfa Aesar Supplier No. H52168.06
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 158171-14-3 | C25H24NO8P | 497.44 g/mol

It can be applied in the synthesis of phosphopeptides. This derivative can be introduced using standard activation methods, such as PyBOP and TBTU. The monoprotected phosphoserine residue once incorporated is stable to piperidine. Using this reagent, even peptides containing multiple phosphorylation sites can be prepared efficiently by standard Fmoc SPPS methods. β-piperidinylalanine formation has been shown to occur during Fmoc deprotection of N-terminal Ser(PO(OBzl)OH), particularly under microwave conditions. This side reaction can be eliminated by using cyclohexylamine or DBU just for this Fmoc deprotection step .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be applied in the synthesis of phosphopeptides. This derivative can be introduced using standard activation methods, such as PyBOP and TBTU. The monoprotected phosphoserine residue once incorporated is stable to piperidine. Using this reagent, even peptides containing multiple phosphorylation sites can be prepared efficiently by standard Fmoc SPPS methods. β-piperidinylalanine formation has been shown to occur during Fmoc deprotection of N-terminal Ser(PO(OBzl)OH), particularly under microwave conditions. This side reaction can be eliminated by using cyclohexylamine or DBU just for this Fmoc deprotection step .

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158171-14-3
Molecular Formula C25H24NO8P
Molecular Weight (g/mol) 497.44
MDL Number MFCD00797869
InChI Key ZBPUWGDUVAAWJY-QHCPKHFHSA-N
Synonym fmoc-o-benzylphospho-l-serine, fmoc-ser hpo3bzl-oh, fmoc-o-benzyl-l-phosphoserine, fmoc-ser po3bzlh-oh, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzyloxy hydroxy phosphoryl oxy propanoic acid, 2s-3-benzyloxy hydroxy phosphoryl oxy-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, pubchem20516, fmoc-l-ser po obzl oh-oh, n-fmoc-o-benzylphospho-l-serine, fmoc-ser po3bzlh-oh hplc
PubChem CID 11005563
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[hydroxy(phenylmethoxy)phosphoryl]oxypropanoic acid
SMILES C1=CC=C(C=C1)COP(=O)(O)OCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 5 g
Beilstein 7060483
Solubility Information Slightly soluble in water.
Formula Weight 497.44
Percent Purity 95%
Chemical Name or Material O-Benzylphospho-N-Fmoc-L-serine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.