Learn More
O-Benzylhydroxylamine hydrochloride, 99%, Thermo Scientific Chemicals
£18.60 - £156.00
Chemical Identifiers
CAS | 2687-43-6 |
---|---|
Molecular Formula | C7H10ClNO |
Molecular Weight (g/mol) | 159.613 |
MDL Number | MFCD00012952 |
InChI Key | HYDZPXNVHXJHBG-UHFFFAOYSA-N |
Synonym | o-benzylhydroxylamine hydrochloride, benzyloxyamine hydrochloride, o-benzylhydroxylamine hcl, o-benzyl-hydroxylamine hydrochloride, hydroxylamine, o-phenylmethyl-, hydrochloride, hydroxylamine, o-benzyl-, hydrochloride, o-benzyl-hydroxylamine hcl, o-benzyl hydroxylamine hydrochloride, aminooxy methyl benzene hydrochloride, oxybenzylamine, chloride |
PubChem CID | 102312 |
IUPAC Name | O-benzylhydroxylamine;hydrochloride |
SMILES | C1=CC=C(C=C1)CON.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11473637
|
Thermo Scientific Alfa Aesar
A17393.06 |
5 g |
£18.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11483637
|
Thermo Scientific Alfa Aesar
A17393.14 |
25 g |
£54.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493637
|
Thermo Scientific Alfa Aesar
A17393.22 |
100 g |
£156.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For use in the formation of nitrones from carbonyl compounds and their use in cycloaddition reactions. Used as a reagent used in the synthesis of hydroxylamines and hydroxyamates. Effective reagent used to prepare α-hydroxybenzylamines from α-hydroxyketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFor use in the formation of nitrones from carbonyl compounds and their use in cycloaddition reactions. Used as a reagent used in the synthesis of hydroxylamines and hydroxyamates. Effective reagent used to prepare α-hydroxybenzylamines from α-hydroxyketones.
Solubility
Soluble in DMSO and methanol.
Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
Chemical Identifiers
2687-43-6 | |
159.613 | |
HYDZPXNVHXJHBG-UHFFFAOYSA-N | |
102312 | |
C1=CC=C(C=C1)CON.Cl |
C7H10ClNO | |
MFCD00012952 | |
o-benzylhydroxylamine hydrochloride, benzyloxyamine hydrochloride, o-benzylhydroxylamine hcl, o-benzyl-hydroxylamine hydrochloride, hydroxylamine, o-phenylmethyl-, hydrochloride, hydroxylamine, o-benzyl-, hydrochloride, o-benzyl-hydroxylamine hcl, o-benzyl hydroxylamine hydrochloride, aminooxy methyl benzene hydrochloride, oxybenzylamine, chloride | |
O-benzylhydroxylamine;hydrochloride |
Specifications
2687-43-6 | |
C7H10ClNO | |
3595440 | |
o-benzylhydroxylamine hydrochloride, benzyloxyamine hydrochloride, o-benzylhydroxylamine hcl, o-benzyl-hydroxylamine hydrochloride, hydroxylamine, o-phenylmethyl-, hydrochloride, hydroxylamine, o-benzyl-, hydrochloride, o-benzyl-hydroxylamine hcl, o-benzyl hydroxylamine hydrochloride, aminooxy methyl benzene hydrochloride, oxybenzylamine, chloride | |
HYDZPXNVHXJHBG-UHFFFAOYSA-N | |
O-benzylhydroxylamine;hydrochloride | |
102312 | |
99% |
∼238°C (sublimation) | |
MFCD00012952 | |
Hygroscopic | |
Soluble in DMSO and methanol. | |
C1=CC=C(C=C1)CON.Cl | |
159.613 | |
159.62 | |
O-Benzylhydroxylamine hydrochloride |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 220-249-0
missing translation for 'rtecsNumber' : NC3040000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only