missing translation for 'onlineSavingsMsg'
Learn More

O-Benzylhydroxylamine, 97%

Product Code. 10270113 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10270113 25 g Glass bottle 25g
10795983 5 g Glass bottle 5g
10592744 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10270113 Supplier Thermo Scientific Acros Supplier No. 382050250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-33-3 | C7H9NO | 123.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-33-3
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.61
MDL Number MFCD00221709
InChI Key HYDZPXNVHXJHBG-UHFFFAOYSA-N
Synonym benzyloxyamine, hydroxylamine, o-phenylmethyl, hydroxylamine, o-benzyl, o-benzyl-hydroxylamine, benzyl-o-hydroxylamine, oxybenzylamine, o-phenylmethyl hydroxylamine, o-benzyloxylamine, phenylmethoxyamine, benzyloxy amine
PubChem CID 102313
IUPAC Name hydrogen O-benzylhydroxylamine chloride
SMILES [H+].[Cl-].NOCC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.1900g/mL
Boiling Point 110°C to 112°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2ONH2
Refractive Index 1.5390 to 1.5410 (20°C, 589nm)
Quantity 25 g
Beilstein 06, IV, 2562
Specific Gravity 1.19
Solubility Information Solubility in water: soluble
Formula Weight 123.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material O-Benzylhydroxylamine
Show More Show Less
compliance-icons
Product Identifier
  • O-Benzylhydroxylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.