missing translation for 'onlineSavingsMsg'
Learn More

O-Benzyl-L-tyrosine benzyl ester hydrochloride, 95%, Thermo Scientific™

Catalog Number 15468156
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
52142-01-5
C23H24ClNO3
397.90
MFCD00153465
DYLKTBYVTWFYGQ-DKVUPULANA-N
h-tyr bzl-obzl.hcl, h-tyr bzl-obzl hcl, o-benzyl-l-tyrosine benzyl ester hydrochloride, l-tyrosine, o-phenylmethyl-, phenylmethyl ester, hydrochloride, httyr bzl-obzl hcl, h-tyr bzl-obzlcl, h-tyr bzl-obzl??hcl, h-tyr bzl-obzl hydrochloride, benzyl o-benzyl-l-tyrosinate-hydrogen chloride 1/1, benzyl 2s-2-amino-3-4-benzyloxyphenyl propanoate hydrochloride
45073233
benzyl (2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoate;hydrochloride
Cl.N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(=O)OCC1=CC=CC=C1
This item is not returnable. View return policy
52142-01-5
C23H24ClNO3
397.90
MFCD00153465
DYLKTBYVTWFYGQ-DKVUPULANA-N
h-tyr bzl-obzl.hcl, h-tyr bzl-obzl hcl, o-benzyl-l-tyrosine benzyl ester hydrochloride, l-tyrosine, o-phenylmethyl-, phenylmethyl ester, hydrochloride, httyr bzl-obzl hcl, h-tyr bzl-obzlcl, h-tyr bzl-obzl??hcl, h-tyr bzl-obzl hydrochloride, benzyl o-benzyl-l-tyrosinate-hydrogen chloride 1/1, benzyl 2s-2-amino-3-4-benzyloxyphenyl propanoate hydrochloride
45073233
benzyl (2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoate;hydrochloride
Cl.N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(=O)OCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 397.89
Percent Purity 95%
Quantity 1 g
Physical Form Solid
Chemical Name or Material O-Benzyl-L-tyrosine benzyl ester hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'