missing translation for 'onlineSavingsMsg'
Learn More

o-Anisidine, 99%

Product Code. 11444023
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11444023

Brand: Thermo Scientific Alfa Aesar A10288.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90-04-0 | C7H9NO | 123.155 g/mol

o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.

Solubility
Soluble in dilute inorganic acid, alcohol, diethyl ether. Not miscible in water.

Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents, acid anhydrides, chloroformates, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-04-0
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00007688
InChI Key VMPITZXILSNTON-UHFFFAOYSA-N
Synonym o-anisidine, 2-anisidine, ortho-anisidine, 2-aminoanisole, o-methoxyaniline, o-aminoanisole, benzenamine, 2-methoxy, 2-methoxybenzenamine, o-methoxyphenylamine, o-anisylamine
PubChem CID 7000
ChEBI CHEBI:82288
IUPAC Name 2-methoxyaniline
SMILES COC1=CC=CC=C1N

Specifications

Melting Point 5°C to 6°C
Density 1.092
Boiling Point 224°C to 225°C
Flash Point 98°C (208°F)
Assay Percent Range 99%
Refractive Index 1.574
Quantity 1000 g
UN Number UN2431
Beilstein 386210
Sensitivity Light sensitive
Merck Index 14,667
Solubility Information Soluble in dilute inorganic acid,alcohol,diethyl ether. Not miscible in water.
Formula Weight 123.16
Percent Purity 99%
Chemical Name or Material o-Anisidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • o-Anisidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : o-Anisidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.