missing translation for 'onlineSavingsMsg'
Learn More

o-Anisidine, 99%, Thermo Scientific Chemicals

Catalog Number 11454023
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
90-04-0
C7H9NO
123.155
MFCD00007688
VMPITZXILSNTON-UHFFFAOYSA-N
o-anisidine, 2-anisidine, ortho-anisidine, 2-aminoanisole, o-methoxyaniline, o-aminoanisole, benzenamine, 2-methoxy, 2-methoxybenzenamine, o-methoxyphenylamine, o-anisylamine
7000
CHEBI:82288
2-methoxyaniline
COC1=CC=CC=C1N
This item is not returnable. View return policy
90-04-0
C7H9NO
123.155
MFCD00007688
VMPITZXILSNTON-UHFFFAOYSA-N
o-anisidine, 2-anisidine, ortho-anisidine, 2-aminoanisole, o-methoxyaniline, o-aminoanisole, benzenamine, 2-methoxy, 2-methoxybenzenamine, o-methoxyphenylamine, o-anisylamine
7000
CHEBI:82288
2-methoxyaniline
COC1=CC=CC=C1N

o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Anisidine is used in dyes, perfume and pharmaceutical intermediate. It is used to produce azo dye, azoic dye.

Solubility
Soluble in dilute inorganic acid, alcohol, diethyl ether. Not miscible in water.

Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents, acid anhydrides, chloroformates, acids.
TRUSTED_SUSTAINABILITY
Melting Point 5°C to 6°C
Density 1.092 g/mL
Boiling Point 224°C to 225°C
Flash Point 98°C (208°F)
Assay Percent Range 99%
Refractive Index 1.574
Quantity 250 g
UN Number UN2431
Beilstein 386210
Sensitivity Light sensitive
Merck Index 14,667
Solubility Information Soluble in dilute inorganic acid,alcohol,diethyl ether. Not miscible in water.
Formula Weight 123.16
Percent Purity 99%
Chemical Name or Material o-Anisidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'