missing translation for 'onlineSavingsMsg'
Learn More

O-Acetylserine, MedChemExpress

Product Code. 16444596
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16444596

Brand: Medchem Express HY101409

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress O-Acetylserine (O-Acetyl-L-serine) is an intermediate in the biosynthesis of the amino acid cysteine in bacteria and plants.

O-Acetylserine (O-Acetyl-L-serine) is an intermediate in the biosynthesis of the amino acid cysteine in bacteria and plants.

Specifications

Chemical Name or Material O-Acetylserine
Physical Form Solid
CAS 5147-00-2
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C5H9NO4
Synonym O-Acetyl-L-serine
Solubility Information H2O : 50 mg/mL (339.84 mM; Need ultrasonic) ∣DMSO : 6.25 mg/mL (42.48 mM; ultrasonic and warming and heat to 60°C)
SMILES N[C@@H](COC(C)=O)C(O)=O
Molecular Weight (g/mol) 147.13
Formula Weight 147.13
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.