missing translation for 'onlineSavingsMsg'
Learn More

O-Acetylsalicyloyl chloride, 97%

Product Code. p-7043472
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11364364

Brand: Thermo Scientific Alfa Aesar L00919.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5538-51-2 | C9H7ClO3 | 198.602 g/mol

O-Acetylsalicyloyl chloride was used in the synthesis of enantiomerically pure bidentate heteroorganic ligands built on simple achiral skeletons and containing an aziridine moiety. It was used in the general synthesis of acetoxybenzamides. It was used in the chemical synthesis of 2,2,6,6-tetramethyl-1-piperidinyloxy(TEMPO)-aspirin conjugate via condensation reaction with 4-hydroxy-TEMPO. It was used as reagent in acylation of cyclobutenediones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Acetylsalicyloyl chloride was used in the synthesis of enantiomerically pure bidentate heteroorganic ligands built on simple achiral skeletons and containing an aziridine moiety. It was used in the general synthesis of acetoxybenzamides. It was used in the chemical synthesis of 2,2,6,6-tetramethyl-1-piperidinyloxy(TEMPO)-aspirin conjugate via condensation reaction with 4-hydroxy-TEMPO. It was used as reagent in acylation of cyclobutenediones.

Solubility
Soluble in toluene. Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Store away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5538-51-2
Molecular Formula C9H7ClO3
Molecular Weight (g/mol) 198.602
MDL Number MFCD00000663
InChI Key DSGKWFGEUBCEIE-UHFFFAOYSA-N
Synonym 2-acetoxybenzoyl chloride, o-acetylsalicyloyl chloride, acetylsalicyloyl chloride, 2-chlorocarbonyl phenyl acetate, benzoyl chloride, 2-acetyloxy, aspirin chloride, o-acetylsalicyloylchloride, acetylsalicoyl chloride, 2-carbonochloridoyl phenyl acetate, ascc
PubChem CID 79668
IUPAC Name (2-carbonochloridoylphenyl) acetate
SMILES CC(=O)OC1=CC=CC=C1C(=O)Cl

Specifications

Melting Point 47°C to 50°C
Boiling Point 134°C to 136°C (12 mmHg)
Flash Point >110°C (230°F)
Odor Pungent, Makes Eyes Water
Quantity 5 g
UN Number UN3261
Beilstein 880372
Formula Weight 198.61
Percent Purity 97%
Chemical Name or Material O-Acetylsalicyloyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.