missing translation for 'onlineSavingsMsg'
Learn More

O-Acetylsalicyloyl chloride, 97%

Product Code. 11364364
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364364 5 g 5g
11374364 25 g 25g
11384364 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11364364 Supplier Thermo Scientific Alfa Aesar Supplier No. L00919.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5538-51-2 | C9H7ClO3 | 198.602 g/mol

O-Acetylsalicyloyl chloride was used in the synthesis of enantiomerically pure bidentate heteroorganic ligands built on simple achiral skeletons and containing an aziridine moiety. It was used in the general synthesis of acetoxybenzamides. It was used in the chemical synthesis of 2,2,6,6-tetramethyl-1-piperidinyloxy(TEMPO)-aspirin conjugate via condensation reaction with 4-hydroxy-TEMPO. It was used as reagent in acylation of cyclobutenediones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Acetylsalicyloyl chloride was used in the synthesis of enantiomerically pure bidentate heteroorganic ligands built on simple achiral skeletons and containing an aziridine moiety. It was used in the general synthesis of acetoxybenzamides. It was used in the chemical synthesis of 2,2,6,6-tetramethyl-1-piperidinyloxy(TEMPO)-aspirin conjugate via condensation reaction with 4-hydroxy-TEMPO. It was used as reagent in acylation of cyclobutenediones.

Solubility
Soluble in toluene. Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Store away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5538-51-2
Molecular Formula C9H7ClO3
Molecular Weight (g/mol) 198.602
MDL Number MFCD00000663
InChI Key DSGKWFGEUBCEIE-UHFFFAOYSA-N
Synonym 2-acetoxybenzoyl chloride, o-acetylsalicyloyl chloride, acetylsalicyloyl chloride, 2-chlorocarbonyl phenyl acetate, benzoyl chloride, 2-acetyloxy, aspirin chloride, o-acetylsalicyloylchloride, acetylsalicoyl chloride, 2-carbonochloridoyl phenyl acetate, ascc
PubChem CID 79668
IUPAC Name (2-carbonochloridoylphenyl) acetate
SMILES CC(=O)OC1=CC=CC=C1C(=O)Cl

Specifications

Melting Point 47°C to 50°C
Boiling Point 134°C to 136°C (12 mmHg)
Flash Point >110°C (230°F)
Odor Pungent, Makes Eyes Water
Quantity 5 g
UN Number UN3261
Beilstein 880372
Formula Weight 198.61
Percent Purity 97%
Chemical Name or Material O-Acetylsalicyloyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.