missing translation for 'onlineSavingsMsg'
Learn More

O-Acetylsalicylic acid, 99%

Product Code. 11464114
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11464114

Brand: Thermo Scientific Alfa Aesar A12488.0E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50-78-2 | C9H8O4 | 180.16 g/mol

O-Acetylsalicylic acid is the prototypical analgesic used in the treatment of mild to moderate pain. acts as an inhibitor of cyclooxygenase which results in the inhibition of the biosynthesis of prostaglandins. Aspirin also inhibits platelet aggregation and is used in the prevention of arterial and venous thrombosis. Along with sodium salicylate (sc-3520), aspirin has been shown to block the long terminal repeat from the human immunodeficiency virus and the Ig κ enhancer from NFκB dependent transcription.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Acetylsalicylic acid is the prototypical analgesic used in the treatment of mild to moderate pain. acts as an inhibitor of cyclooxygenase which results in the inhibition of the biosynthesis of prostaglandins. Aspirin also inhibits platelet aggregation and is used in the prevention of arterial and venous thrombosis. Along with sodium salicylate (sc-3520), aspirin has been shown to block the long terminal repeat from the human immunodeficiency virus and the Ig κ enhancer from NFκB dependent transcription.

Solubility
Soluble in 100% ethanol (80mg/ml), DMSO (41mg/ml) or dimethyl formamide (30mg/ml); slightly soluble in Phosphate-buffered saline (PBS), pH 7.2 (2.7mg/ml).

Notes
Store at room temperature. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-78-2
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00002430
InChI Key BSYNRYMUTXBXSQ-UHFFFAOYSA-N
Synonym aspirin, acetylsalicylic acid, 2-acetoxybenzoic acid, 2-acetyloxy benzoic acid, acetylsalicylate, o-acetylsalicylic acid, o-acetoxybenzoic acid, acylpyrin, polopiryna, easprin
PubChem CID 2244
ChEBI CHEBI:15365
IUPAC Name 2-acetyloxybenzoic acid
SMILES CC(=O)OC1=CC=CC=C1C(O)=O

Specifications

Melting Point 133°C to 140°C
Density 1.4
Flash Point 250°C (482°F)
Quantity 2500 g
Beilstein 779271
Merck Index 14,851
Solubility Information Soluble in 100% ethanol (80mg/ml),DMSO (41mg/ml) or dimethyl formamide (30mg/ml); slightly soluble in Phosphate-buffered saline (PBS),pH 7.2 (2.7mg/ml).
Formula Weight 180.15
Percent Purity 99%
Chemical Name or Material O-Acetylsalicylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.